Skip to Content
Merck
All Photos(1)

Key Documents

M6065

Sigma-Aldrich

Monoclonal Anti-Microphthalmia antibody produced in mouse

clone C5, purified immunoglobulin

Synonym(s):

Anti-Mi

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€635.00

€635.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1335


Select a Size

Change View
100 μG
€635.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€635.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1335

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C5, monoclonal

form

buffered aqueous solution

mol wt

antigen 52-56 kDa

species reactivity

mouse, rat, human

concentration

0.5-1.0 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable using 2 μg/mg protein lysate
western blot: 1:500 using 10 μg of Mouse brain lysates

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MITF(4286)
mouse ... Mitf(17342)
rat ... Mitf(25094)

General description

Microphthalmia (Mi in mouse or MITF in human) is a basic helix-loop-helix-leucine zipper (BHLH–ZIP) transcription factor involved in the differentiation, development and survival of melanocytes and cells of the retinal pigment epithelium, i.e. cells responsible for hair, skin, and eye color. It activates the expression of the melanocyte specific genes tyrosinase and TRP1 (tyrosinase-related protein 1) by binding as a homo- or heterodimer to a symmetrical DNA sequence (E box) (5′-CATGTG-3′) located within the M box found in their promoters. Microphthalmia also appears to be involved in the differentiation of mast cells, osteoclasts, basophils and natural killer cells.
Microphthalmia is expressed in a limited number of cell types including heart, mast cells, osteoclast precursors, and melanocytes. There are a number of different isoforms of microphthalmia resulting from alternative splicing and alternative promotors. These isoforms differ at their amino-termini and in their expression patterns.
Mouse monoclonal clone C5 anti-Microphthalmia antibody recognizes serine phosphorylated and non-phosphorylated melanocytic isoforms of microphthalmia from human, mouse or rat.

Immunogen

N-terminal fragment of human microphthalmia protein.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Mouse monoclonal clone C5 anti-Microphthalmia antibody is used to tag serine phosphorylated and non-phosphorylated melanocytic isoforms of microphthalmia for detection and quantitation by techniques such as immunoblotting (doublet of 52-56 kDa), immunoprecipitation, immunohistochemistry on formalin-fixed paraffin embedded or frozen tissue sections, and gel shift. It is used as a probe to determine the roles of microphthalmia in the differentiation, development and survival of melanocytes and cells of the retinal pigment epithelium.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Pieper et al.
Oncoimmunology, 7(8), e1450127-e1450127 (2018-09-18)
The profound but frequently transient clinical responses to BRAFV600 inhibitor (BRAFi) treatment in melanoma emphasize the need for combinatorial therapies. Multiple clinical trials combining BRAFi and immunotherapy are under way to further enhance therapeutic responses. However, to which extent BRAFV600
Xueping Wang et al.
PloS one, 10(11), e0143142-e0143142 (2015-11-19)
TYR, DCT and MITF are three important genes involved in maintaining the mature phenotype and producing melanin; they therefore participate in neural crest cell development into melanocytes. Previous studies have revealed that the Wnt signaling factor lymphoid enhancer-binding factor (LEF-1)
Jared Iacovelli et al.
Investigative ophthalmology & visual science, 57(3), 1038-1051 (2016-03-11)
Oxidative stress and metabolic dysregulation of the RPE have been implicated in AMD; however, the molecular regulation of RPE metabolism remains unclear. The transcriptional coactivator, peroxisome proliferator-activated receptor-gamma coactivator 1α (PGC-1α) is a powerful mediator of mitochondrial function. This study
Li-Ping Liu et al.
Cell reports, 27(2), 455-466 (2019-04-11)
Induced pluripotent stem cells (iPSCs) are a promising melanocyte source as they propagate indefinitely and can be established from patients. However, the in vivo functions of human iPSC-derived melanocytes (hiMels) remain unknown. Here, we generated hiMels from vitiligo patients using a three-dimensional
Li-Ping Liu et al.
STAR protocols, 1(1), 100004-100004 (2020-10-29)
Melanocytes, derived from neural crest cells, are involved in melanin production. This protocol describes a method to generate induced melanocytes (iMelanocytes) from human induced pluripotent stem cells (iPSCs) using a suspension culture system, which considerably improves the differentiation efficiency. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service