Skip to Content
Merck
All Photos(1)

Key Documents

M5814

Sigma-Aldrich

MKK6/SKK3, active human

≥80% (SDS-PAGE), recombinant, expressed in E. coli, Maltose binding protein tagged, buffered aqueous solution

Synonym(s):

MAP Kinase Kinase 6, MAP Kinase Kinase 6/SKK 3

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€682.00

€682.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
€682.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€682.00


Estimated to ship on20 May 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

81 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP2K6(5608)

Specificity

Active in the phosphorylation of p38/SAPK2a

Application

Active enzyme for study of stress-activated protein kinases.
May be used for activation of p38α followed by phosphorylation of MBP.

Unit Definition

1 unit will activate 1 unit of p38/SAPK2 per minute. 1 unit of active p38/SAPK2 will incorporate 1 nanomole of phosphorus into myelin basic protein per minute.

Physical form

Solution in approx. 1 mg/ml in 50 mM Tris-HCl, pH 7.5, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.15 M NaCl, 0.03% Brij® L23, 270 mM sucrose, 0.2 mM PMSF, 1 mM benzamidine.

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Cuenda et al.
The EMBO journal, 16(2), 295-305 (1997-01-15)
Stress-activated protein kinase-3 (SAPK3), a recently described MAP kinase family member with a wide-spread tissue distribution, was transfected into several mammalian cell lines and shown to be activated in response to cellular stresses, interleukin-1 (IL-1) and tumour necrosis factor (TNF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service