Skip to Content
Merck
All Photos(1)

Key Documents

M5269

Sigma-Aldrich

Moniliformin sodium salt from Fusarium proliferatum

Synonym(s):

1-Hydroxycyclobut-1-ene-3,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€488.00

€488.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€488.00

About This Item

Linear Formula:
C4HO3Na
CAS Number:
Molecular Weight:
120.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€488.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

storage temp.

2-8°C

SMILES string

O=C1C(C=C1[O-])=O.[Na+]

InChI

1S/C4H2O3.Na/c5-2-1-3(6)4(2)7;/h1,5H;/q;+1/p-1

InChI key

FERDNJVXTWPNSA-UHFFFAOYSA-M

General description

Moniliformin (MON), a mycotoxin[1] and small ionic molecule[2], is present in various Fusarium species including Fusarium avenaceum, Fusarium subglutinans and Fusarium proliferatum.[1] It is present as sodium or potassium salt of semisquaric acid naturally.[1] MON is also present in maize and small-grain cereals.[2]

Application

Moniliformin sodium salt from Fusarium proliferatum has been used as a mycotoxin standard:
  • to test its acute oral toxicity in mice[1]
  • to test its subacute toxic effects in rats[2]
  • in characterizing mycotoxins from Aspergillus[3]

Biochem/physiol Actions

Moniliformin (MON) is implicated for its toxic potential and may lead to respiratory distress and progressive muscular weakness in rats.[1] It inhibits the tricarboxylic acid (TCA) cycle oxidation step.[1] By acting as a pyruvate substrate, MON effectively inhibits thiamine pyrophosphate cofactor dependant enzymes and blocks the gluconeogenesis pathway.[1] ) Furthermore, MON also inhibits glutathione peroxidase and glutathione reductase leading to oxidative stress in myoblast.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H R Burmeister et al.
Applied and environmental microbiology, 37(1), 11-13 (1979-01-01)
Fusarium moniliforme NRRL 6322 produced about 600 mg of recoverable moniliformin, a mycotoxic metabolite, per kg of corn grit medium. The moniliformin was extracted from the grits with methanol, purified by preparative thin-layer chromatography, and crystallized from ether. The 50%
Fusarium species colonizing spears and forming mycotoxins in field samples of asparagus from Germany and Poland
Weber Z, et al.
Journal of Phytopathology, 209-216 null
Martina Jonsson et al.
Toxicology letters, 233(1), 38-44 (2014-12-09)
Moniliformin is a Fusarium mycotoxin mainly produced by several species infecting grains in different climatic conditions. According to our previous studies, it is acutely toxic to rats, with an LD50 cut-off value of 25mg/kg b.w. To further assess the possible
Katharina Walburga von Bargen et al.
Journal of agricultural and food chemistry, 60(14), 3586-3591 (2012-03-21)
Moniliformin is a mycotoxin produced by fungi of the Fusarium genus and occurs as a contaminant of different cereals worldwide. This study describes the first application of isotopically labeled (13)C(2)-moniliformin for the analysis of moniliformin in cereals. Moniliformin is a
An Zhang et al.
Journal of Zhejiang University. Science. B, 11(3), 200-208 (2010-03-06)
To investigate the effects of mycotoxin moniliformin (MON) on the metabolism of aggrecan and type II collagen in human chondrocytes in vitro and the relationship between MON and Kashin-Beck disease (KBD). Human chondrocytes were isolated and cultured on bone matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service