Skip to Content
Merck
All Photos(1)

Key Documents

M5196

Sigma-Aldrich

Monoclonal Anti-Macrophage Inflammatory Protein-2 antibody produced in rat

clone 40605, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CXCL2, Anti-Chemokine (C-X-C motif) ligand 2, Anti-GROb, Anti-MIP2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

40605, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: 2 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Cxcl2(20310)

Immunogen

purified, E. coli-derived recombinant mouse macrophage inflammatory protein 2.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service