Skip to Content
Merck
All Photos(1)

Key Documents

M4824

Sigma-Aldrich

Myosin Light Chain from human heart

Synonym(s):

Myosin from human heart

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€493.00

€493.00


Check Cart for Availability


Select a Size

Change View
100 μG
€493.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61

€493.00


Check Cart for Availability

biological source

human heart

Quality Level

form

lyophilized

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from ammonium carbonate buffer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service