Skip to Content
Merck

Skip To

M4686

Greiner high and medium binding 96 well plates

flat bottom white wells, high binding surface, sterile

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates, Greiner 655074

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
40 ea
Check Cart for Availability
€ 176,00

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom white wells, white polystyrene
Size:
96 wells
Sterility:
sterile
Binding type:
high binding surface
Feature:
lid: no, skirt (F-bottom)

€ 176,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom white wells, white polystyrene

sterility

sterile

feature

lid: no, skirt (F-bottom)

packaging

case of 40

manufacturer/tradename

Greiner 655074

maximum volume

392 μL

size

96 wells

working volume

340 μL

binding type

high binding surface

General description

LUMITRAC 600 96W Microplate, well working volume 25-340 μL; total well volume 392 μL.
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Application

White plates minimize cross talk in luminescence procedures. LUMITRAC 600 will bind up to 600 ng/cm2. Suitable for cell work. Free of detectable DNase/RNase, human DNA and pyrogens

Other Notes

96 well flat (F) bottom; high binding microplate, without lid, Chimney Style; alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Legal Information

LUMITRAC is a trademark of Greiner Bio-One GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5686M4936M5811
feature

lid: no, skirt (F-bottom)

feature

lid: no, skirt (F-bottom)

feature

lid: no, skirt (F-bottom)

feature

lid: no, skirt (F-bottom)

material

flat bottom white wells, white polystyrene

material

black polystyrene plate, flat bottom clear polystyrene wells (with micro-clear bottom)

material

black polystyrene , flat bottom black wells

material

black polystyrene , flat bottom clear polystyrene wells (with micro-clear bottom)

binding type

high binding surface

binding type

medium binding surface

binding type

medium binding surface

binding type

high binding surface

manufacturer/tradename

Greiner 655074

manufacturer/tradename

Greiner 655096

manufacturer/tradename

Greiner 655076

manufacturer/tradename

Greiner 655097

sterility

sterile

sterility

non-sterile

sterility

non-sterile

sterility

sterile

working volume

340 μL

working volume

340 μL

working volume

340 μL

working volume

340 μL


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library