Skip to Content
Merck

Skip To

M3668

Metergoline

Serotonin antagonist

Synonym(s):

N-CBZ-[(8β)-1,6-Dimethylergolin-8-yl]methylamine, [(8β)-1,6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 163,00
5 g
Check Cart for Availability
€ 1.490,00

About This Item

Empirical Formula (Hill Notation):
C25H29N3O2
CAS Number:
Molecular Weight:
403.52
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
241-686-3
MDL number:

€ 163,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Metergoline,

Quality Segment

mp

148-150 °C (lit.)

solubility

0.1 M HCl: 1.4 mg/mL, ethanol: 4 mg/mL, H2O: insoluble

storage temp.

−20°C

SMILES string

[H][C@]2(CNC(=O)OCc1ccccc1)CN(C)[C@]3([H])Cc4cn(C)c5cccc(c45)[C@@]3([H])C2

InChI

1S/C25H29N3O2/c1-27-14-18(13-26-25(29)30-16-17-7-4-3-5-8-17)11-21-20-9-6-10-22-24(20)19(12-23(21)27)15-28(22)2/h3-10,15,18,21,23H,11-14,16H2,1-2H3,(H,26,29)/t18-,21+,23+/m0/s1

InChI key

WZHJKEUHNJHDLS-QTGUNEKASA-N

Application

Metergoline has been used as a serotonin receptor antagonist:

  • to study its effects on astrocyte calcium signals evoked by whisker stimulation[1]
  • to study its effects on lipopolysaccharide-induced anorexia in rats[2]
  • to evaluate the non-specific binding by 5-HT1A receptor binding assays[3]

Biochem/physiol Actions

Metergoline is an ergot alkaloid[4] and a selective serotonin antagonist which blocks the 5-HT2A and 5-HT2C receptors with higher affinity.[5] It possesses antifungal activity against infection caused by Candida krusei.[6] Metergoline exhibits a therapeutic effect against premenstrual dysphoric disorder. It also exhibits antipyretic and analgesic activities.[7]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U103T5202B9935
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

solubility

0.1 M HCl: 1.4 mg/mL, H2O: insoluble, ethanol: 4 mg/mL

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 10 mg/mL, 0.1 M HCl: >40 mg/mL, ethanol: >40 mg/mL, 0.1 M NaOH: insoluble, H2O: insoluble

solubility

DMSO: >5 mg/mL

solubility

DMSO: 18 mg/mL, H2O: insoluble

mp

148-150 °C (lit.)

mp

-

mp

-

mp

-

Gene Information

human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR7(3363)
rat ... Htr7(65032)

Gene Information

human ... OPRD1(4985), OPRK1(4986), OPRM1(4988)
mouse ... Oprk1(18387)
rat ... Oprd1(24613), Oprk1(29335), Oprm1(25601)

Gene Information

-

Gene Information

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters