Skip to Content
Merck
All Photos(1)

Key Documents

M3074

Sigma-Aldrich

MMPIP

≥98% (HPLC)

Synonym(s):

6-(4-Methoxyphenyl)-5-methyl-3-(4-pyridinyl)-isoxazolo[ 4,5-c]pyridin-4(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N3O3
CAS Number:
Molecular Weight:
333.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥10 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

room temp

SMILES string

COc1ccc(cc1)C2=Cc3onc(-c4ccncc4)c3C(=O)N2C

InChI

1S/C19H15N3O3/c1-22-15(12-3-5-14(24-2)6-4-12)11-16-17(19(22)23)18(21-25-16)13-7-9-20-10-8-13/h3-11H,1-2H3

InChI key

PDWYBOZNEVALOV-UHFFFAOYSA-N

Biochem/physiol Actions

MMPIP is a potent, selective, allosteric antagonist of metabotropic glutamate receptor 7 (mGluR7). It also displays intrinsic activity and acts as an inverse agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Du et al.
Nature, 594(7864), 589-593 (2021-06-18)
The metabotropic glutamate receptors (mGlus) are involved in the modulation of synaptic transmission and neuronal excitability in the central nervous system1. These receptors probably exist as both homo- and heterodimers that have unique pharmacological and functional properties2-4. Here we report
Paulina Cieślik et al.
Frontiers in molecular neuroscience, 11, 316-316 (2018-10-09)
The data concerning antipsychotic-like activity of negative allosteric modulators (NAMs)/antagonists of mGlu7 receptors are limited. The only available ligands for this receptor are MMPIP and ADX71743. In the present studies, we used stable cell line expressing mGlu7 receptor and it
George A Lemieux et al.
PLoS biology, 11(11), e1001712-e1001712 (2013-11-22)
Phenotypic screens can identify molecules that are at once penetrant and active on the integrated circuitry of a whole cell or organism. These advantages are offset by the need to identify the targets underlying the phenotypes. Additionally, logistical considerations limit
Jolanta H Kotlinska et al.
European journal of pharmacology, 858, 172512-172512 (2019-07-02)
The present study was conducted to evaluate the influence of AMN082, the metabotropic glutamate receptor subtype 7 (mGlu7) allosteric agonist on different stages of memory processes connected with fear conditioning in the passive avoidance (PA) learning task in mice and
Sinead E Shortall et al.
Molecular neurobiology, 57(8), 3439-3457 (2020-06-14)
Despite several compounds entering clinical trials for the negative and cognitive symptoms of schizophrenia, few have progressed beyond phase III. This is partly attributed to a need for improved preclinical models, to understand disease and enable predictive evaluation of novel

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service