Skip to Content
Merck
All Photos(1)

Key Documents

M120

Sigma-Aldrich

Metaphit methanesulfonate salt

≥98% (HPLC), solid

Synonym(s):

1-[1-(3-Isothiocyanato)phenyl]cyclohexylpiperidine methanesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H24N2S · CH3SO3H
CAS Number:
Molecular Weight:
396.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

H2O: >20 mg/mL

storage temp.

−20°C

SMILES string

CS(O)(=O)=O.S=C=Nc1cccc(c1)C2(CCCCC2)N3CCCCC3

InChI

1S/C18H24N2S.CH4O3S/c21-15-19-17-9-7-8-16(14-17)18(10-3-1-4-11-18)20-12-5-2-6-13-20;1-5(2,3)4/h7-9,14H,1-6,10-13H2;1H3,(H,2,3,4)

InChI key

ZASHKPXYYPYQRI-UHFFFAOYSA-N

Gene Information

human ... OPRS1(10280)

Biochem/physiol Actions

Metaphit methanesulfonate salt is an irreversible, non-competitive antagonist at the phencyclidine site on the NMDA receptor; sigma receptor irreversible, competitive antagonist.

Caution

Hygroscopic

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L S Bluth et al.
European journal of pharmacology, 161(2-3), 273-277 (1989-02-28)
Pretreatment of guinea pig brain membranes with 1-[1-(3-isothiocyanatophenyl)cyclohexyl]piperidine (Metaphit) caused irreversible and differential inhibition of ligand binding to sigma (sigma) receptors. The concentration of Metaphit required to produce 50% inhibition of binding of [3H]1,3-di-o-tolylguanidine ([3H]DTG), [3H](+)-3-(3-hydroxy-phenyl)-N-(1-propyl)piperidine ([3H](+)-3-PPP), and [3H](+)-N-allylnormetazocine ([3H](+)-SKF
I Zimányi et al.
Synapse (New York, N.Y.), 3(3), 239-245 (1989-01-01)
[3H]Mazindol was used to label the dopamine uptake complex in mouse striatum in vitro in the presence and absence of metaphit, an isothiocyanate derivative of phencyclidine. In some experiments, metaphit was present in the incubation fluid throughout the procedure; in
E D French et al.
European journal of pharmacology, 140(3), 267-274 (1987-08-21)
Metaphit, a derivative of phencyclidine (PCP) which irreversibly binds to a population of PCP receptor sites in rat brain, has been considered as a potentially specific PCP antagonist. In whole animal studies, however, metaphit has been shown to have either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service