Skip to Content
Merck
All Photos(1)

Key Documents

M045030

Sigma-Aldrich

6-Tetrachloro-Fluorescein Phosphoramidite

configured for ABI, configured for ÄKTA® and OligoPilot®

Synonym(s):

6-(4, 7, 2′, 7′-Tetrachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MMOL
€317.00

€317.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 MMOL
€317.00

About This Item

Empirical Formula (Hill Notation):
C46H54N3O10Cl4P
Molecular Weight:
981.72
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

€317.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (31P-NMR)
≥97.0% (reversed phase HPLC)

color

white to off-white, powder

suitability

conforms to structure for H-NMR
conforms to structure for H-NMR

compatibility

configured for ABI
configured for ÄKTA® and OligoPilot®

storage temp.

−70°C

SMILES string

O=C(C1=C(Cl)C=C(C(NCCCCCCOP(N(C(C)C)C(C)C)OCCC#N)=O)C(Cl)=C12)OC32C4=C(C=C(OC(C(C)(C)C)=O)C(Cl)=C4)OC5=CC(OC(C(C)(C)C)=O)=C(Cl)C=C53

InChI

1S/C46H54Cl4N3O10P/c1-25(2)53(26(3)4)64(59-19-15-16-51)58-18-14-12-11-13-17-52-40(54)27-20-32(49)37-38(39(27)50)46(63-41(37)55)28-21-30(47)35(61-42(56)44(5,6)7)23-33(28)60-34-24-36(31(48)22-29(34)46)62-43(57)45(8,9)10/h20-26H,11-15,17-19H2,1-10H3,(H,52,54)

InChI key

RKVIHIDUTMZQEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Yu et al.
PloS one, 4(7), e6445-e6445 (2009-08-04)
Stringently controlled conditional expressing systems are crucial for the functional characterization of genes. Currently, screening of multiple clones to identify the tightly controlled ones is necessary but time-consuming. Here, we describe a system fusing Tet (tetracycline)-inducible elements, BAC (bacterial artificial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service