L9234
LB Broth (Lennox)
Liquid microbial growth medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Recommended Products
grade
Molecular Biology
Quality Level
sterility
sterile; 0.2 μm filtered
form
liquid
suitability
suitable for nonselective for Escherichia coli and Coliforms
storage temp.
room temp
General description
Liquid media is also termed as broth. It has nutrients dissolved in water. Luria-Bertani broth (LB broth) is used in naphthalene degradation experiments.
Application
LB Broth has been used to culture P. aeruginosa.
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added.
Biochem/physiol Actions
LB Lennox media is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates.
Features and Benefits
LB Lennox media provides:
- Filter-sterilized, ready-to-use format
- Convenient package size
- Standard formulation
Components
10 g/L Tryptone
5.0 g/L Yeast Extract
5.0 g/L NaCl
5.0 g/L Yeast Extract
5.0 g/L NaCl
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pseudomonas toxin pyocyanin triggers autophagy: Implications for pathoadaptive mutations
Yang ZS, et al.
Autophagy, 12(6), 1015-1028 (2016)
Understanding Microbes: A Laboratory Textbook for Microbiology (1989)
Su-Yun Chang et al.
Water environment research : a research publication of the Water Environment Federation, 87(1), 61-67 (2015-01-30)
Luria-Bertani broth and acetone were usually used in naphthalene degradation experiments as nutrient and solvent. However, their effect on the degradation was seldom mentioned. In this work, we investigated the effect of LB, naphthalene concentration, and acetone on the degradation
Protocols
Initiating a Starter Culture
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service