Skip to Content
Merck
All Photos(1)

Key Documents

L9000

Sigma-Aldrich

L-Leucine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
181.66
Beilstein:
3595133
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

151-153 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].COC(=O)[C@@H](N)CC(C)C

InChI

1S/C7H15NO2.ClH/c1-5(2)4-6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H/t6-;/m0./s1

InChI key

DODCBMODXGJOKD-RGMNGODLSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Brown et al.
Nature, 380(6572), 345-347 (1996-03-28)
The prion protein PrPc is a glycoprotein of unknown function normally found in neurons and glia. It is involved in diseases such as bovine spongiform encephalopathy (BSE), scrapie and Creutzfeldt-Jakob disease. PrPSc, an altered isoform of PrPC that is associated
M G Leonardi et al.
Archives of insect biochemistry and physiology, 48(4), 190-198 (2001-12-18)
Nutrient absorption and its modulation are critical for animal growth. In this paper, we demonstrate that leucine methyl ester (Leu-OMe) can greatly increase the activity of the transport system responsible for the absorption of most essential amino acids in the
Holly H Birdsall et al.
Journal of immunology (Baltimore, Md. : 1950), 175(5), 3347-3353 (2005-08-24)
One hundred ten to 120-kDa fragments of fibronectin (FNf), generated by proteases released in the course of tissue injury and inflammation, stimulate monocytes to produce proinflammatory cytokines, promote mononuclear leukocytes (MNL) transendothelial migration, up-regulate monocyte CD11b and CD86 expression, and
C Fraisier et al.
Journal of immunological methods, 176(1), 9-22 (1994-11-10)
Peripheral blood lymphocytes from healthy HIV-1 seronegative donors were immunized in vitro with the following synthetic peptides: (i) an octameric poly-L-lysine conjugated peptide of the HIV-1MN V3 loop and (ii) a resin bound synthetic peptide aa642-665 of HIV-1 gp41. Lymphoblastoid
Mary E Hamby et al.
Journal of neuroscience methods, 150(1), 128-137 (2005-08-18)
Cultures of astrocytes can be readily established and are widely used to study the biological functions of these glial cells in isolation. Unfortunately, contamination by microglia can confound results from such studies. Herein, a simple and highly effective modification of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service