Skip to Content
Merck
All Photos(1)

Key Documents

L8411

Sigma-Aldrich

Ultima Gold XR LSC Cocktail

For aqueous samples

Synonym(s):

Liquid Scintillation Counting, Scintillation Cocktail

Sign Into View Organizational & Contract Pricing

Select a Size

2 X 5 L
€1,580.00

€1,580.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
2 X 5 L
€1,580.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

€1,580.00


Estimated to ship on23 May 2025


Request a Bulk Order

General description

Ultima Gold XR LSC Cocktail is a safer line of liquid scintillation cocktails (LSC) designed for a wide range of aqueous solutions. Liquid scintillation counting in radionuclide metrology involves preparing samples which are emulsions of aqueous radioactive solutions in liquid scintillation cocktails.[1] Ultima Gold  XR LSC Cocktail technology is an alternative screening tool to alpha spectrometry and gas flow proportional counting.

Application

Ultima Gold XR LSC Cocktail is widely used to measure low-energy radionuclides. Metrology laboratories also use it for the standardization of radioactive solutions.

Features and Benefits

Ultima Gold XR LSC Cocktail has the following benefits:

  • A high flash point (150 °C) offers greater safety.
  • High sample load capacity and high counting efficiency − up to 50% for 3H. 
  • Compatible with alkaline samples.
  • It will not diffuse through polyethylene vials. 
  • Biodegradable. 
  • Compatible with polar organic solvents.

Legal Information

Ultima Gold is a trademark of Revvity

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youcef Nedjadi et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 118, 25-31 (2016-09-03)
The stability of 3H and 63Ni samples, made with Ultima Gold and Ultima Gold AB using eight aqueous fractions ranging between 0.1% and 10%, was monitored over a period of 32 weeks. Counting rate losses were measured in all cases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service