Skip to Content
Merck
All Photos(1)

Key Documents

L7651

Sigma-Aldrich

Lysozyme from chicken egg white

lyophilized powder, protein ≥90 %, ≥40,000 units/mg protein, for molecular biology

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

grade

Molecular Biology
for molecular biology

form

lyophilized powder

specific activity

≥40,000 units/mg protein

mol wt

single-chain 14.3 kDa

composition

protein, ≥90%

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... LYZ(396218)

Looking for similar products? Visit Product Comparison Guide

Application

Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.
Lysozyme hydrolyzes the β-1,4 linkages between N-acetylmuramic acid and N-acetylglucosamine, a polysaccharide backbone of peptidoglycans in the cell wall structure of many microorganisms. This is particularly useful for lysing Gram-positive and Gram-negative bacteria for subsequent nucleic acid extraction.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).
Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan.

Features and Benefits

  • Highly purified by repeated crystallization and dialysis
  • Each lot is use-tested for isolation of plasmid DNA from E. coli

Unit Definition

One unit will produce a ΔA450 of 0.001 per min at pH 6.24 at 25°C, using a suspension of Micrococcus lysodeikticus as substrate, in a 2.6 mL reaction mixture (1 cm light path).

Physical form

Lyophilized powder, essentially salt-free

Preparation Note

3× crystallized

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How can I reconstitute Lysozyme from chicken egg white, Product L7651?

    For E. coli cell lysis, use a freshly prepared lysozyme solution (10 mg/mL) in 10 mM Tris-HCl, pH 8.0. The product is also soluble in water (10 mg/mL) yielding a clear to slightly hazy colorless solution.

  4. How stable are solutions of Lysozyme from chicken egg white, Product L7651?

    Aqueous solutions should retain activity for at least one month when stored between 2-8 °C.

  5. Is Lysozyme from chicken egg white, Product L7651, suitable for bacterial lysis prior to DNA purification?

    Each lot is verified to be suitable for use as a lysing agent in purifying plasmid DNA from E. coli. You can also use this product in combination with the GenElute Bacterial Genomic DNA Kit (Product No. NA2100) to purify genomic DNA from Gram-positive bacteria.

  6. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  7. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  8. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 1-1 (1989)
Lauri Mikonranta et al.
Journal of invertebrate pathology, 144, 88-96 (2017-02-07)
The insect immune system has versatile ways of coping with microbial insults. Currently, innate immune priming has been described in several invertebrates, and the first insights into its mechanistic basis have been described. Here we studied infections with two different
Anuj Shukla et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(13), 5075-5080 (2008-03-26)
In colloidal systems, the interplay between the short range attraction and long-range repulsion can lead to a low density associated state consisting of clusters of individual particles. Recently, such an equilibrium cluster phase was also reported for concentrated solutions of
Sara A Bumgardner et al.
PloS one, 13(5), e0196950-e0196950 (2018-05-08)
Safe and efficacious orally-delivered mucosal vaccine platforms are desperately needed to combat the plethora of mucosally transmitted pathogens. Lactobacillus spp. have emerged as attractive candidates to meet this need and are known to activate the host innate immune response in
Sibel Ebru Yalcin et al.
Nature chemical biology, 16(10), 1136-1142 (2020-08-19)
Multifunctional living materials are attractive due to their powerful ability to self-repair and replicate. However, most natural materials lack electronic functionality. Here we show that an electric field, applied to electricity-producing Geobacter sulfurreducens biofilms, stimulates production of cytochrome OmcZ nanowires

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service