Skip to Content
Merck
All Photos(2)

Key Documents

L6645

Sigma-Aldrich

Lacto-N-difucohexaose II

from human milk, ~95%

Synonym(s):

β-D-Gal-(1→3)-[α-L-Fuc-(1→4)]-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-[α-L-Fuc-(1→3)]-D-Glc, LNH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H65NO29
CAS Number:
Molecular Weight:
999.91
Beilstein:
1340391
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

human milk

Assay

~95%

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@H]([C@@H](O)C=O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]4O)[C@H](O[C@@H]5O[C@H](CO)[C@H](O)[C@H](O)[C@H]5O)[C@H]3NC(C)=O)[C@H]2O)[C@H](O)CO)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C38H65NO29/c1-9-18(48)22(52)25(55)35(59-9)64-29(12(46)4-40)30(13(47)5-41)65-38-28(58)33(21(51)15(7-43)62-38)68-34-17(39-11(3)45)32(67-37-27(57)24(54)20(50)14(6-42)61-37)31(16(8-44)63-34)66-36-26(56)23(53)19(49)10(2)60-36/h4,9-10,12-38,41-44,46-58H,5-8H2,1-3H3,(H,39,45)/t9-,10-,12-,13+,14+,15+,16+,17+,18+,19+,20-,21-,22+,23+,24-,25-,26-,27+,28+,29+,30+,31+,32+,33?,34-,35-,36-,37-,38-/m0/s1

InChI key

QNSWVQWPVUZACH-GFBQVKCGSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Lewis-a trisaccharide linked to 3-fucosyllactose

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Cancilla et al.
Analytical chemistry, 71(15), 3206-3218 (1999-08-18)
A method is described to obtain multicollision dissociation threshold (MCDT) values. These values provide relative reaction thresholds for dissociation in the three major gas-phase fragmentation reactions of oligosaccharides complexed to alkali metal ions. The quasimolecular ions are produced using matrix-assisted
Structural and conformational studies of carbohydrate oncodevelopmental antigens.
E F Hounsell
Annales de l'Institut Pasteur. Immunology, 138(6), 863-867 (1987-11-01)
A S Donald et al.
Carbohydrate research, 178, 79-91 (1988-07-15)
Lacto-N-neo-difucohexaose II, beta-D-Galp-(1----4)-[alpha-L-Fucp-(1----3)]-beta-D-Glcp-NAc-(1----3)-beta-D-Galp-(1----4)-[alpha-L-Fucp-(1----3)]-D-Glc, and 3'-galactosyllactose, beta-D-Galp-(1----3)-beta-D-Galp-(1----4)-D-Glc, were isolated for the first time from human milk by means of a recycling chromatography technique. Through this method, carried out mainly on columns of K+ ion-exchange resins and either Bio-Gel P-4 or TSK
Xi Huang et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(6), 708-717 (2018-10-06)
1. The pharmacokinetics (PKs) analysis of compounds absorbed after the oral administration of Si-Ni-San (SNS) decoction to functional dyspepsia (FD) patients was designed to detect whether the effects were similar to prokinetics administered to healthy rats, without ethical limitation. 2.
M Elwakiel et al.
Journal of agricultural and food chemistry, 66(27), 7036-7043 (2018-06-19)
To study the variability in human milk oligosaccharide (HMO) composition of Chinese human milk over a 20-wk lactation period, HMO profiles of 30 mothers were analyzed using CE-LIF. This study showed that total HMO concentrations in Chinese human milk decreased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service