Skip to Content
Merck
All Photos(1)

Key Documents

L6394

Sigma-Aldrich

Lysozyme human

lyophilized powder, ≥100,000 units/mg protein

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥100,000 units/mg protein

composition

Protein, ~10% E1%/280

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LYZ(4069)

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will produce a ΔA450 of 0.001 per min at pH 6.24 at 25 °C, using a suspension of Micrococcus lysodeikticus as substrate, in a 2.6 mL reaction mixture (1 cm light path).

Physical form

Lyophilized powder containing sodium phosphate and sodium chloride

Preparation Note

Derived from human milk

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P F Zabrodskiĭ et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(11), 19-21 (2013-01-18)
It is established in experiments on noninbred rats that the use of imunofan (20 mg/kg daily) and polyoxidonium (150 mg/kg daily) for 7 days on the background of chronic intoxication with organophosphorus agent VX (0.01 LD50, single daily treatment for
Mariela G Del Giudice et al.
Infection and immunity, 81(3), 956-964 (2013-01-16)
Secretion of proteins in Gram-negative bacteria is a high-energy-consuming process that requires translocation across two membranes and a periplasmic space composed of a mesh-like layer, the peptidoglycan. To achieve this, bacteria have evolved complex secretion systems that cross these barriers
Anne Derbise et al.
The Journal of infectious diseases, 207(10), 1535-1543 (2013-02-14)
Yersinia pestis (the plague bacillus) and its ancestor, Yersinia pseudotuberculosis (which causes self-limited bowel disease), encode putative homologues of the periplasmic lysozyme inhibitor Ivy and the membrane-bound lysozyme inhibitor MliC. The involvement of both inhibitors in virulence remains subject to
Mahdieh Mansouri et al.
Molecules (Basel, Switzerland), 18(1), 789-813 (2013-01-25)
The interactions between cyclophosphamide (CYC) and lysozyme (LYZ) in the presence of different cyclodextrins (CDs) were investigated by UV absorption, fluorescence spectroscopy, circular dichroism (CD), and molecular modeling techniques under imitated physiological conditions. The UV absorption results showed the formation
Zhe Wang et al.
The journal of physical chemistry. B, 117(4), 1186-1195 (2013-01-11)
High-resolution inelastic X-ray scattering was used to investigate the collective vibrational excitations in hydrated lysozyme powders as a function of hydration level and temperature. It is found that the samples with strong enzymatic function are "soft", in the sense that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service