Skip to Content
Merck
All Photos(1)

Documents

L5408

Sigma-Aldrich

LY-171,883

≥98% (TLC)

Synonym(s):

5-[4-(4-Acetyl-3-hydroxy-2-propylphenoxy)butyl]-1H-tetrazole, Tomelukast

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O3
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

SMILES string

CCCc1c(O)c(ccc1OCCCCc2nnn[nH]2)C(C)=O

InChI

1S/C16H22N4O3/c1-3-6-13-14(9-8-12(11(2)21)16(13)22)23-10-5-4-7-15-17-19-20-18-15/h8-9,22H,3-7,10H2,1-2H3,(H,17,18,19,20)

InChI key

MWYHLEQJTQJHSS-UHFFFAOYSA-N

Biochem/physiol Actions

Selective leukotriene D4 (LTD4) receptor antagonist; PPARα and PPARγ agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Peters et al.
Molecular pharmacology, 50(1), 67-74 (1996-07-01)
Peroxisome proliferator-activated receptor alpha (PPAR alpha) mediates the effects of foreign chemical peroxisome proliferators on liver and kidney, including the induction of peroxisomal, mitochondrial, and microsomal enzymes involved in beta-oxidation of fatty acids. However, the role of this receptor in
Kirsten Scholz-Pedretti et al.
Journal of the American Society of Nephrology : JASN, 13(3), 611-620 (2002-02-22)
Natural activators of peroxisome proliferator-activated receptors (PPAR) are lipid metabolites, including those produced by phospholipases A(2) (PLA(2)). In glomerular mesangial cells, the secreted group IIA PLA(2) (sPLA(2)-IIA), which is thought to be a crucial factor in pathologic processes in the
Common structural requirements for peroxisome proliferation by tetrazole and carboxylic acid-containing compounds.
P I Eacho et al.
Annals of the New York Academy of Sciences, 804, 387-402 (1996-12-27)
K Yamakawa et al.
Biochemical and biophysical research communications, 271(3), 571-574 (2000-05-18)
Vascular endothelial growth factor (VEGF), expressed in a variety of mesenchymal cells including vascular smooth muscle cells (VSMC), is a potent mitogen for endothelial cells, and is used clinically applied for ischemic disease of peripheral vessels. To determine whether peroxisome
N L Anderson et al.
Toxicology and applied pharmacology, 137(1), 75-89 (1996-03-01)
We have investigated the effects of five peroxisome proliferators (PPs : clofibric acid, DEHP, WY14,643, nafenopin, and LY171883) on the abundances of a large number of proteins in the livers of treated mice at 5- and 35-day time points. LY171883

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service