Skip to Content
Merck
All Photos(1)

Key Documents

L3910

Millipore

Lowenstein Jensen medium base

BioChemika, suitable for microbiology

Synonym(s):

L. J. Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

application(s)

microbiology

suitability

mycobacteria

Application

Lowenstein Jensen Medium with the addition of eggs is used for cultivation of Mycobacteria, especially Mycobacterium tuberculosis.

Components

Components (g/L)
L-Asparagine 3.60
Monopotassium Phosphate 2.40
Magnesium Sulphate 0.24
Magnesium Citrate 0.60
Potato Flour 30.00
Malachite Green 0.40

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lowenstein, E.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten Und Hygiene. 1. Abt. Medizinisch-Hygienische Bakteriologie, Virusforschung und Parasitologie. Originale, 120, 127-127 (1931)
Jensen, K.A.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten Und Hygiene. 1. Abt. Medizinisch-Hygienische Bakteriologie, Virusforschung und Parasitologie. Originale, 125, 222-222 (1932)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service