Skip to Content
Merck
All Photos(3)

Key Documents

L131

Sigma-Aldrich

L-745,870 hydrochloride

Synonym(s):

3-[[4-(4-Chlorophenyl)piperazin-1-yl]methyl]-1H-pyrrolo[2,3-b]pyridine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€297.00
25 MG
€966.00

€297.00


Estimated to ship on18 May 2025



Select a Size

Change View
5 MG
€297.00
25 MG
€966.00

About This Item

Empirical Formula (Hill Notation):
C18H19ClN4 · HCl
CAS Number:
Molecular Weight:
363.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€297.00


Estimated to ship on18 May 2025


form

solid

Quality Level

color

white

solubility

H2O: 0.2 mg/mL
ethanol: 1.4 mg/mL
0.1 M HCl: 5.5 mg/mL

SMILES string

Cl.Clc1ccc(cc1)N2CCN(CC2)Cc3c[nH]c4ncccc34

InChI

1S/C18H19ClN4.ClH/c19-15-3-5-16(6-4-15)23-10-8-22(9-11-23)13-14-12-21-18-17(14)2-1-7-20-18;/h1-7,12H,8-11,13H2,(H,20,21);1H

InChI key

GFFJYISJZJIKAF-UHFFFAOYSA-N

Gene Information

human ... DRD4(1815)

Biochem/physiol Actions

Selective D4 dopamine receptor antagonist.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Manufactured and sold under license from Merck & Co., Inc., Kenilworth, NJ, U.S. for use solely for preclinical researchy purposes (i.e., not for the administration to or other use in humans).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Patel et al.
The Journal of pharmacology and experimental therapeutics, 283(2), 636-647 (1997-11-14)
L-745,870,(3-([4-(4-chlorophenyl)piperazin-1-yl]methyl)-1H- pyrollo[2,3-b] pyridine, was identified as a selective dopamine D4 receptor antagonist with excellent oral bioavailability and brain penetration. L-745,870 displaced specific binding of 0.2 nM [3H] spiperone to cloned human dopamine D4 receptors with a binding affinity (Ki) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service