L1290
Lactoferricin B, fragment 4-14 trifluoroacetate salt
≥95% (HPLC)
Synonym(s):
bLFC (26-36), bovine lactoferricin
Select a Size
€322.00
Select a Size
About This Item
€322.00
Recommended Products
Quality Level
Assay
≥95% (HPLC)
solubility
H2O: soluble
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
Mode of action
cell membrane | interferes
storage temp.
−20°C
Amino Acid Sequence
General description
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service