Skip to Content
Merck
All Photos(1)

Documents

L1168

Sigma-Aldrich

Anti-phospho-5-Lipoxygenase (pSer523) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ALOX5(240)
rat ... Alox5(25290)

General description

The gene ALOX5 (arachidonate 5-lipoxygenase), also referred to as 5-LOX (5-lipoxygenase), encodes a protein belonging to the lipoxygenase family. The gene is mapped to human chromosome 10q11.21. The protein contains a distinct ‘C2-like′ domain at the amino terminal that is involved in Ca2+-dependent membrane binding and a large catalytic domain containing the non-heme catalytic iron. It is highly unstable with short-lived activity.

Immunogen

phosphopeptide corresponding to amino acid residues surrounding the pSer523 of human 5-lipoxgenase (5-LO).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

The gene ALOX5 (arachidonate 5-lipoxygenase) encodes a lipoxygenase that catalyzes the conversion of arachidonic acid (AA) and other fatty acids to bioactive compounds, such as 5(S)-hydroxyeicosatetraenoic acid (5-HETE), leukotriene B4 (LTB4), and other leukotrienes. These leukotrienes and lipoxins function as mediators of inflammatory response. ALOX5 has been implicated in tumorigenesis and may serve as a potential therapeutic target and biomarker.

Physical form

Solution in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structure of human 5-lipoxygenase.
Gilbert NC
Science, 331, 217-219 (2011)
ALOX5 polymorphism associates with increased leukotriene production and reduced lung function and asthma control in children with poorly controlled asthma.
Mougey E
Clinical and Experimental Allergy, 43, 512-520 (2013)
Fangming Tang et al.
The Journal of experimental medicine, 212(10), 1487-1495 (2015-08-26)
Eicosanoids are inflammatory mediators that play a key but incompletely understood role in linking the innate and adaptive immune systems. Here, we show that cytotoxic effector T cells (CTLs) are capable of both producing and responding to cysteinyl leukotrienes (CystLTs)
Arachidonate 5 lipoxygenase expression in papillary thyroid carcinoma promotes invasion via MMP-9 induction.
Kummer NT
Journal of Cellular Biochemistry, 113, 1998-2008 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service