Skip to Content
Merck
All Photos(1)

Key Documents

K4389

Sigma-Aldrich

KR-32568

≥98% (HPLC), solid

Synonym(s):

[5-(2-Methyl-5-fluorophenyl)furan-2-ylcarbonyl]guanidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€230.00
25 MG
€689.00

€230.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€230.00
25 MG
€689.00

About This Item

Empirical Formula (Hill Notation):
C13H12FN3O2
CAS Number:
Molecular Weight:
261.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€230.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ~10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc(F)cc1-c2ccc(o2)C(=O)NC(N)=N

InChI

1S/C13H12FN3O2/c1-7-2-3-8(14)6-9(7)10-4-5-11(19-10)12(18)17-13(15)16/h2-6H,1H3,(H4,15,16,17,18)

InChI key

DTLDHYBZLVASJQ-UHFFFAOYSA-N

Gene Information

human ... SLC9A1(6548)

Biochem/physiol Actions

Sodium/hydrogen exchanger-1 (NHE-1) inhibitor.
Sodium/hydrogen exchanger-1 (NHE-1) inhibitor; IC50 = 0.23 μM; inhibited NHE-1-mediated rabbit platelet swelling; in anesthetized rats, reduced infarct size from 67% (control) to 43% (at 0.1 mg/kg) and 24% (at 1.0 mg/kg); reduced number of ventricular premature beats from 530 to 266 (at 0.1 mg/kg) and 115 (at 1.0 mg/kg); reduced ventricular fibrillation incidence from 17 to 8 (0.1 mg/kg) and 0 (1.0 mg/kg); implications for research and treatment of myocardial ischemic diseases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniele Torella et al.
Journal of the American Heart Association, 3(2), e000434-e000434 (2014-03-29)
Diabetes mellitus (DM) has multifactorial detrimental effects on myocardial tissue. Recently, carbonic anhydrases (CAs) have been shown to play a major role in diabetic microangiopathy but their role in the diabetic cardiomyopathy is still unknown. We obtained left ventricular samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service