Skip to Content
Merck

K1376

β-Ketoglutaric acid

Synonym(s):

1,3-Acetonedicarboxylic acid, 3-Oxopentanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
208-797-9
UNSPSC Code:
12352103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library