Skip to Content
Merck
All Photos(1)

Key Documents

K1005

Sigma-Aldrich

Monoclonal Anti-Kinesin antibody produced in mouse

clone IBII, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

IBII, monoclonal

mol wt

antigen 40-80 kDa (Two bands. Additional bands at 130-140 kDa may be present.)

contains

15 mM sodium azide

species reactivity

rat, sea urchin, frog, bovine

technique(s)

western blot: 1:500 using kinesin-enriched rat brain preparation

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Specificity

The antibody recognizes the kinesin heavy chain.

Immunogen

kinesin from bovine brain.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greta Forlani et al.
The FEBS journal, 273(10), 2127-2138 (2006-05-03)
RasGRF is a family of guanine nucleotide exchange factors with dual specificity for both Ras and Rac GTPases. In this study, using mouse brain extracts, we show that both RasGRF1 and RasGRF2 interact with microtubules in an in vitro microtubule
Nicholas Angeloni et al.
The journal of sexual medicine, 10(5), 1240-1250 (2012-09-22)
The cavernous nerve (CN) is commonly injured during prostatectomy, resulting in erectile dysfunction (ED). Although peripheral nerves have a limited ability to regenerate, a return of function typically does not occur due to irreversible down stream morphological changes in the
Rollin W Robinson et al.
Histochemistry and cell biology, 122(1), 1-5 (2004-06-09)
Using a new fixation solution, CytoSkelFix, it is now possible to obtain superior fixation and thus resolution of cytoskeletal components using immunofluorescence and fluorescence microscopy. This fixative combines rapid cell penetration and cellular crosslinking of proteins such that both preservation
G Amadoro et al.
Neurobiology of aging, 32(6), 969-990 (2009-07-25)
Alzheimer's disease (AD) is characterized by Aβ overproduction and tau hyperphosphorylation. We report that an early, transient and site-specific AD-like tau hyperphosphorylation at Ser262 and Thr231 epitopes is temporally and causally related with an activation of the endogenous amyloidogenic pathway
Yatma Gueye et al.
Glia, 59(5), 750-770 (2011-03-02)
Olfactory ensheathing cells (OECs) are unique glia found only in the olfactory system. They retain exceptional plasticity and support olfactory neurogenesis and retargeting across the PNS:CNS boundary in the olfactory system. OECs have been shown to improve functional outcome when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service