Skip to Content
Merck
All Photos(3)

Key Documents

K0265

Sigma-Aldrich

Anti-Potassium Channel K2P4.1 (TRAAK) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCNK4, Anti-TWIK-related arachidonic acid-stimulated K+ channel

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€307.00

€307.00


Estimated to ship on29 May 2025



Select a Size

Change View
50 μL
€307.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€307.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: 1:200 using HEK-K2P4.1 transfected cells or rat cerebellum lysate

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNK4(50801)
rat ... Kcnk4(116489)

General description

The gene K2P4.1, also referred to as KCNK4 (potassium two pore domain channel subfamily K member 4), encodes a protein belonging to the subfamily of K2P (two pore domain K+ channel) channels called TRAAK (TWIK-related arachidonic acid-stimulated K+ channel). The TRAAK channels are expressed selectively in the nervous system. TRAAK channel contains two protomers, each with two distinct pore domains forming a two-fold symmetric potassium channel.

Specificity

The K2P4.1 antibody is directed against a highly conserved epitope in the intraracellular C-terminal region of the human K2P4.1 channel.

Immunogen

peptide NLAFIDESSDTQSERGC, corresponding to amino acid residues 343-359 of human K2P4.1.

Biochem/physiol Actions

The gene KCNK4 (potassium two pore domain channel subfamily K member 4) encodes a potassium channel that is mechanosensitive and is regulated by polyunsaturated fatty acids and temperature. The structure and chemical composition of the lipid bilayer has a profound effect on the functioning of this K+ channel. It is found to regulate the noxious input threshold in dorsal root ganglia neurons.

Physical form

Lyophilized powder from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein, such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of the human K2P TRAAK, a lipid- and mechano-sensitive K+ ion channel.
Brohawn SG
Science, 335, 436-441 (2012)
Trichloroethanol enhances the activity of recombinant human TREK-1 and TRAAK channels.
Harinath S and Sikdar SK
Neuropharmacology, 46, 750-760 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service