Skip to Content
Merck
All Photos(1)

Documents

J4127

Sigma-Aldrich

Anti-Jagged 1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-AGS, Anti-AGS1, Anti-AHD, Anti-AWS, Anti-CD339, Anti-CMT2HH, Anti-DCHE, Anti-HJ1, Anti-JAGL1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

capture ELISA: 0.2-0.8 μg/mL
flow cytometry: 2.5 μg using 106 rat cortical stem cells
western blot: 0.1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JAG1(182)
rat ... Jag1(29146)

General description

Notch signaling is a well conserved among the metazoans and is a regulator of growth, differentiation, and patterning processes. One of the ligands of Notch family receptors is Jagged. Mammalian Jagged family comprises of Jagged-1 and Jagged-2. Jagged proteins are expressed during the critical stages of embryonic and postnatal development. The Notch-Jagged signaling regulates the proliferation of endothelial cells, apoptosis, differentiation of hematopoietic progenitors, skeletal development and formation and differentiation of blood vessels. Notch-jagged pathway components play important role in the pathology of diseases like Alagille syndrome, CADASIL (cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy) and osteosarcoma. Anti-Jagged 1 antibody specifically recognizes rat Jagged-1.

Immunogen

recombinant rat Jagged 1 extracellular domain, expressed in NSO cells.

Application

Anti-Jagged 1 antibody may be used for ELISA at a working antibody concentration of 0.5-1.0 μg/ml. For immunoblotting, antibody concentration of 0.1-0.2 μg/ml is recommended. For flow cytometry, a concentration of 20 μg/ml (10 μL/105 cells to 106 cells) may be used. The antibody is suitable for neutralization assays (ND50 is ~2-10 μg/ml).

Target description

Jagged-1 is a Notch ligand. The Notch signaling system regulates proliferation, differentiation, and apoptosis in many tissues, including hematopoietic tissue. Notch is a family of transmembrane receptors activated by ligands (Jagged-1, Jagged-2, and Delta-1) that are expressed on adjacent cells. Hematopoietic stem cells and early progenitors express Notch-1 and Notch-2, whereas primary stromal cultures have been shown to express Jagged-1, indicating that Jagged-1 may play a role in nurturing progenitor cells within the hematopoietic microenvironment. Cells engineered to express high levels of Jagged-1 caused a four-fold increase in formation of hematopoietic progenitor cell colonies. However, without the presence of other cytokines that stimulate proliferation and maturation, such as IL-3, IL-6, SCF and G-CSF, Jagged-1 actually decreased myeloid colony formation of CD34+ cells, suggesting that Jagged-1 modulates cytokine signals and helps preserve CD34+ cells in an immature state.

Physical form

lyophilized from a 0.2 μm filtered solution of phosphate buffered saline (PBS), pH 7.4, and 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandramouli R Chillakuri et al.
Seminars in cell & developmental biology, 23(4), 421-428 (2012-02-14)
The Notch receptor is part of a core signalling pathway which is highly conserved in all metazoan species. It is required for various cell fate decisions at multiple stages of development and in the adult organism, with dysregulation of the
Stefano Zanotti et al.
Molecular and cellular biology, 30(4), 886-896 (2009-12-10)
Notch receptors are transmembrane receptors that regulate cell fate decisions. There are four Notch receptors in mammals. Upon binding to members of the Delta and Jagged family of transmembrane proteins, Notch is cleaved and the Notch intracellular domain (NICD) is
Cristina Roca et al.
Genes & development, 21(20), 2511-2524 (2007-10-17)
The Notch pathway is a versatile regulator of cell fate specification, growth, differentiation, and patterning processes in metazoan organisms. In the vertebrate cardiovascular system, multiple Notch family receptors and several of their Jagged and Delta-like ligands are expressed during critical
Ana-Rita Pedrosa et al.
The Prostate, 76(1), 80-96 (2015-10-01)
The Notch signaling pathway has been implicated in prostate development, maintenance and tumorigenesis by its key role in cell-fate determination, differentiation and proliferation. Therefore, we proposed to analyze Notch family members transcription and expression, including ligands (Dll1, 3, 4 and
A V Shah et al.
Nature communications, 8, 16002-16002 (2017-07-12)
Notch and Angiopoietin-1 (Ang1)/Tie2 pathways are crucial for vascular maturation and stability. Here we identify the transcription factor ERG as a key regulator of endothelial Notch signalling. We show that ERG controls the balance between Notch ligands by driving Delta-like

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service