Skip to Content
Merck
All Photos(1)

Key Documents

J3955

Sigma-Aldrich

JTC-801

≥98% (HPLC)

Synonym(s):

N-(4-amino-2-methylquinolin-6-yl)-2-(4-ethylphenoxymethyl)benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€124.00
25 MG
€352.00

€124.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€124.00
25 MG
€352.00

About This Item

Empirical Formula (Hill Notation):
C26H25N3O2 · HCl
CAS Number:
Molecular Weight:
447.96
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€124.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

tan

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CCc1ccc(OCc2ccccc2C(=O)Nc3ccc4nc(C)cc(N)c4c3)cc1

InChI

1S/C26H25N3O2.ClH/c1-3-18-8-11-21(12-9-18)31-16-19-6-4-5-7-22(19)26(30)29-20-10-13-25-23(15-20)24(27)14-17(2)28-25;/h4-15H,3,16H2,1-2H3,(H2,27,28)(H,29,30);1H

InChI key

NQLIYKXNAXKMBL-UHFFFAOYSA-N

Biochem/physiol Actions

JTC-801 is a nociceptin/orphanin FQ peptide (NOP) receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiichi Hiramoto et al.
Photodermatology, photoimmunology & photomedicine, 33(2), 84-91 (2017-01-01)
We previously reported that ultraviolet (UV) A eye irradiation reduces the ulcerative colitis induced by dextran sodium sulfate (DSS). This study examined the effects of UVA on colon carcinoma induced by azoxymethane (AOM) and DSS. We irradiated the eyes of
G Delaney et al.
Journal of neuroendocrinology, 24(12), 1527-1541 (2012-07-28)
Central nociceptin/orphanin FQ (N/OFQ)-expressing neurones are abundantly expressed in the hypothalamus and limbic system and are implicated in the regulation of activity of the hypothalamic-pituitary-adrenal axis (HPA) and stress responses. We investigated the role of the endogenous N/OFQ receptor (NOP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service