Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C17H15F6N5O
CAS Number:
Molecular Weight:
419.32
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
assay
≥98% (HPLC)
form
powder
color
white to off-white
solubility
DMSO: >20 mg/mL
originator
Johnson & Johnson
storage temp.
−20°C
SMILES string
FC(F)(F)c1c(nccc1)N2CCN(CC2)C(=O)Nc3ncc(cc3)C(F)(F)F
InChI
1S/C17H15F6N5O/c18-16(19,20)11-3-4-13(25-10-11)26-15(29)28-8-6-27(7-9-28)14-12(17(21,22)23)2-1-5-24-14/h1-5,10H,6-9H2,(H,25,26,29)
InChI key
JFRYYGVYCWYIDQ-UHFFFAOYSA-N
Biochem/physiol Actions
JNJ 17203212 is a reversible, competitive and potent TRPV1 antagonist that does not inhibit related TRP channels such as TRPV2, TRPV4, or TRPA1. Listed in BJP with IC50= 65 nM. TRPV1 channels are activated by heat, capsaicin and the endocannabinoid anandamide. Interest in TRPV1 antagonists in the transmission and modulation of pain.
JNJ 17203212 is a reversible, competitive and potent TRPV1 antagonist.
Features and Benefits
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
