Skip to Content
Merck
All Photos(1)

Key Documents

J1625

Sigma-Aldrich

Jenner′s stain

Powder

Synonym(s):

Eosin−Methylene Blue

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€207.00

€207.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€207.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

€207.00


Estimated to ship on19 May 2025


Request a Bulk Order

Product Name

Jenner′s stain, suitable for blood stain

form

powder

Quality Level

color

dark green to very dark green

solubility

methanol: 1 mg/mL, blue to very deep blue

suitability

suitable for blood stain

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service