Skip to Content
Merck
All Photos(1)

Documents

I9781

Sigma-Aldrich

Importin β1 human

≥80% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal histidine tagged), buffered aqueous glycerol solution

Synonym(s):

Importin β, Karyopherin β1, p97

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli (N-terminal histidine tagged)

Assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~97 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KPNB1(3837)

General description

Importin β1 has 19 tandemly repeated HEAT (huntingtin, elongation factor 3 (EF3), protein phosphatase 2A (PP2A) and the yeast kinase TOR1) motifs. The gene is mapped to human chromosome 17q21. The protein shuttles between the cytoplasm and nucleus. It belongs to the importin β family of proteins.

Biochem/physiol Actions

Importin β, an import receptor, is a prototype of the nuclear transport receptor family, which comprises in human more than 20 proteins (90-180 kDa). These proteins interact directly with nuclear pore complex (NPC) and mediate nucleocytoplasmic transport. Importin β imports into the nucleus proteins carrying canonical nuclear localization signals (NLS) as well as UsnRNAs, which it binds via adaptor proteins, importin α and snurpotin-1 respectively. Importin β also binds directly, without adaptors, ribosomal proteins, Smad proteins, and virus derived proteins such as HIV Rev and Tat, that contain non classical NLS. In addition, Improtin b has been shown to be involved in the nuclear envelope assembly process.
The mechanism of importin β action in nuclear import can be demonstrated by the well-studied import of proteins containing classical NLS. Importin β forms a complex with Importin α, which, in turn, binds the cargo protein via its NLS. The Impβ/Impα/cargo complex translocates into the nucleus. When the complex reaches the nuclear site of the NPC, Ran-GTP binds the Impβ to form Impβ/Ran-GTP complex and released the Impα and the cargo protein. The Impβ/Ran-GTP complex is then exported to the cytoplasm where the complex dissociates upon hydrolysis of GTP to GDP, making Impβ ready for a new import cycle.

Physical form

Solution in 5% glycerol containing 20 mM Hepes, pH 7.5, 110 mM potassium-acetate, 2 mM magnesium-acetate, 0.5 mM EGTA, 0.1 mM ATP, 2 mM DTT, and protease inhibitors.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian An et al.
International journal of cancer, 103(2), 194-204 (2002-11-28)
Suppression subtractive hybridization (SSH) was applied to identify differentially expressed genes in the SV40LT immortalized human bronchial epithelial cell line Y-BE, with normal human bronchial epithelial cells (HBEC) as a control. Two cDNA libraries of up- and downregulated genes were
G Cingolani et al.
Nature, 399(6733), 221-229 (1999-06-03)
Cytosolic proteins bearing a classical nuclear localization signal enter the nucleus bound to a heterodimer of importin-alpha and importin-beta (also called karyopherin-alpha and -beta). The formation of this heterodimer involves the importin-beta-binding (IBB) domain of importin-alpha, a highly basic amino-terminal
Amnon Harel et al.
Molecular cell, 16(3), 319-330 (2004-11-05)
Importin beta, once thought to be exclusively a nuclear transport receptor, is emerging as a global regulator of diverse cellular functions. Importin beta acts positively in multiple interphase roles: in nuclear import, as a chaperone for highly charged nuclear proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service