Skip to Content
Merck
All Photos(1)

Key Documents

I7284

Sigma-Aldrich

Anti-Interleukin-7 Receptor α/CD127 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CD127, Anti-Il7r

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

flow cytometry: 3.0-10.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il7r(16197)

General description

Interleukin-7 (IL-7) is a stromal-cell derived cytokine that is important in the development of adaptive immunity and maintenance of T cells and B cells. The cognate receptor that binds IL-7 is the IL-7 receptor complex, made up of IL-7 R α (CD127) chain and a γ chain. The expression of IL-7 R is restricted to immature B cells, mature T cells and thymocytes. The binding of IL-7 to its receptor triggers a variety of tightly regulated signaling pathways that are critical for cell survival, proliferation, differentiation and uptake of glucose. The signaling pathways that participate in mediating the effects of IL-7 R are JAK/STAT and PI3K. Additionally, the effect of IL-7 on cell cycle is due to the modulation of proteins p27kip1 and Cdc25A. Reports implicate IL-7 R in glucose metabolism and cell size as it modulates the effects of GLUT4. Anti-interleukin-7 receptor α/CD127 recognizes mouse IL-7 Rα. This antibody shows less than 1% cross reactivity with rhIL-7 Rα.

Immunogen

purified, NSO cell-derived recombinant mouse interleukin-7 soluble receptor α.

Application

Anti-interleukin-7 receptor α/CD127 antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For flow cytometry, the recommended concentration is 3-10 μg/ml/6 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joao T Barata et al.
The Journal of experimental medicine, 200(5), 659-669 (2004-09-09)
Interleukin (IL)-7 is essential for normal T cell development. Previously, we have shown that IL-7 increases viability and proliferation of T cell acute lymphoblastic leukemia (T-ALL) cells by up-regulating Bcl-2 and down-regulating the cyclin-dependent kinase inhibitor p27kip1. Here, we examined
C Pallard et al.
Immunity, 10(5), 525-535 (1999-06-15)
Here, we define the IL-7R-activated signal that promotes survival and proliferation of T cell progenitors and demonstrate that it is distinct from the signals that induce differentiation. We show that IL-7 activates PKB and STAT5 in human thymocytes. Into T
C Kittipatarin et al.
Cell cycle (Georgetown, Tex.), 5(9), 907-912 (2006-04-22)
A balance between survival and proliferative signals maintains a constant number of T lymphocytes that populate the mammalian immune system, a process termed "homeostasis". Central to this process is the availability of a stromal cell product--the cytokine interleukin-7 (IL-7). We
Christina Kittipatarin et al.
Cytokine, 39(1), 75-83 (2007-09-05)
The signaling processes that maintain the homeostatic proliferation of peripheral T-cells and result in their self-renewal largely remain to be elucidated. Much focus has been placed on the anti-apoptotic function of the cytokine, interleukin-7 (IL-7), during T-cell development. But a
Wen Qing Li et al.
The Journal of experimental medicine, 203(3), 573-582 (2006-02-24)
Interleukin (IL)-7 is required for survival and homeostatic proliferation of T lymphocytes. The survival effect of IL-7 is primarily through regulation of Bcl-2 family members; however, the proliferative mechanism is unclear. It has not been determined whether the IL-7 receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service