Skip to Content
Merck
All Photos(1)

Key Documents

I7003

Sigma-Aldrich

Ibogaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O · HCl
CAS Number:
Molecular Weight:
346.89
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

H2O: soluble
methanol: soluble

application(s)

forensics and toxicology

SMILES string

Cl.CCC1CC2CC3C1N(CCc4c3[nH]c5ccc(OC)cc45)C2

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

CNS stimulant and hallucinogen; anti-convulsant.
Ibogaine is a central nervous system stimulant and hallucinogen that has been reported to be a σ2 agonist, a noncompetitive NMDA receptor antagonist at the phencyclidine site and an enhancer of purinergic muscle contractions. It is also a competitive inhibitor of serotonin and dopamine transport. It is an anti-convulsant and has anti-addictive properties against cocaine and heroin.

Preparation Note

Alkaloid isolated from the African shrub, Tabernanthe iboga.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Itzhak et al.
Annals of the New York Academy of Sciences, 844, 245-251 (1998-07-21)
Although the alkaloid ibogaine is a potent hallucinogenic agent some indications suggest that it may be useful for the treatment of opioid and cocaine addiction. The neurochemical mechanism(s) underlying ibogaine effects remain unclear. In the present study we investigated the
M K Mundey et al.
British journal of pharmacology, 129(8), 1561-1568 (2000-04-26)
Ibogaine and 18-methoxycoronaridine are naturally occurring alkaloids reported to possess antiaddictive properties in several models of drug dependence. We have examined their effect at mu-opioid receptors regulating neurogenic contractions of several smooth muscle preparations and also against spontaneous contractions of
S D Glick et al.
Annals of the New York Academy of Sciences, 909, 88-103 (2000-07-27)
Ibogaine, one of several alkaloids found in the root bark of the African shrub Tabernanthe iboga, has been claimed to be effective in treating multiple forms of drug abuse. Problems associated with side effects of ibogaine have spawned a search
K K Szumlinski et al.
Pharmacology, biochemistry, and behavior, 63(3), 457-464 (1999-07-27)
Pretreatment (19 h) with the putative antiaddictive agent, ibogaine, has been shown previously to potentiate cocaine-induced locomotion in rats. The present study demonstrates that the magnitude of this effect of ibogaine is dependent on the previous cocaine history of the
G B Wells et al.
Brain research bulletin, 48(6), 641-647 (1999-07-01)
Ibogaine has been shown to affect biogenic amine levels in selected brain regions. Because of the involvement of these neurotransmitters in drug addiction, the effects of ibogaine on biogenic amine transport may contribute to the potential anti-addictive properties of ibogaine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service