Skip to Content
Merck
All Photos(2)

Key Documents

I6510

Sigma-Aldrich

Monoclonal Anti-Human IgE antibody produced in mouse

Monoclonal Anti-Human IgE antibody produced in mouse
1 of 1 reviewers received a sample product or took part in a promotion

clone GE-1, ascites fluid

Synonym(s):

Monoclonal Anti-Human IgE

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
€357.00
0.25 ML
€712.00

€357.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
0.1 ML
€357.00
0.25 ML
€712.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€357.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

GE-1, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:5,000

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds human IgE; does not bind human IgG, IgA or IgM.
Monoclonal Anti-Human IgE (mouse IgG2b isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulin E (IgE) antibodies are tetramers with two light chains (κ or λ) and two ε-heavy connected by numerous intrachain disulfide bonds. IgE exists in two isoforms, one is associated with the B cell receptor and the other being secreted by plasma cell. It has a short life span in plasma when compared to receptor bounded IgE.

Specificity

Monoclonal Anti-Human IgE is immunospecific for human IgE. No cross-reactivity with human IgG, IgM or purified light chains is observed.

Application

To confirm that a chimaeric monoclonal antibody had episolon chain, an elisa was performed using monoclonal anti-human IgE antibody (clone GE1) at a 1:1000 dilution.

Biochem/physiol Actions

Immunoglobulin E (IgE) is implicated in acute allergic reactions and chronic inflammatory allergic diseases, such as allergic asthma and chronic urticaria (CU). Receptor bounded IgE aids antigen uptake and modulates the differentiation and proliferation of B cells. It confers immunity against helminthic parasites. IgE is also involved in facilitating type I hypersensitivity reactions. It regulates mast cell homeostasis and also exhibits immunomodulatory effects.

Physical form

The product is provided as ascites fluid with 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Analysis Note

The antibody is tested for reactivity against myeloma IgE and allergy-elevated IgE.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessy Elst et al.
Methods in molecular biology (Clifton, N.J.), 2163, 213-218 (2020-08-09)
The basis of traditional flow cytometry allergy diagnosis is measurement of the expression of basophilic surface activation and/or degranulation markers. Basophils, upon encounter with a specific allergen that cross-links surface FcRI-bound IgE antibodies, not only secrete and release quantifiable bioactive
S Nullens et al.
Cytometry. Part B, Clinical cytometry, 84(3), 173-178 (2013-03-02)
Despite the efficiency of venom immunotherapy, the effects on basophils and mast cells remain incompletely understood and probably vary according to the treatment phase. To study the effect of build-up and maintenance venom immunotherapy on individual basophils. Intracellular histamine and
Jielu Liu et al.
Scientific reports, 12(1), 18924-18924 (2022-11-08)
Asthma is a common respiratory disease associated with airway hyperresponsiveness (AHR), airway inflammation and mast cell (MC) accumulation in the lung. Monensin, an ionophoric antibiotic, has been shown to induce apoptosis of human MCs. The aim of this study was
Hans C Oettgen
The Journal of allergy and clinical immunology, 137(6), 1631-1645 (2016-06-07)
Fifty years ago, after a long search, IgE emerged as the circulating factor responsible for triggering allergic reactions. Its extremely low concentration in plasma created significant hurdles for scientists working to reveal its identity. We now know that IgE levels
Arnau Navinés-Ferrer et al.
Journal of immunology research, 2016, 8163803-8163803 (2017-01-18)
IgE is an immunoglobulin that plays a central role in acute allergic reactions and chronic inflammatory allergic diseases. The development of a drug able to neutralize this antibody represents a breakthrough in the treatment of inflammatory pathologies with a probable

Questions

  1. Could I know the approximate yield of this ascitic liquid (purification by affinity)?

    1 answer
    1. The concentration of antibody in this ascitic fluid is typically in the range of 6 to 10 mg/mL.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service