Skip to Content
Merck
All Photos(1)

Documents

I4907

Sigma-Aldrich

Anti-IKKε/IKK-i, C-Terminal antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IKB Kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1 μg/mL

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IKBKE(9641)

General description

IKK-epsilon/IKK-I belongs to IκB kinase complex and express mainly in immune cells. IKK-I express in response to proinflammatory cytokines such as tumor necrosis factor-α, IL-, IL-6 and stimulate NF-κB activation, hence referred as inducible IκB kinase . Polyclonal anti-IKK epsilon/IKK-I c-terminal antibody can be used in immunoblotting to detect human IKK epsilon/IKK-I. Anti-IKK epsilon/IKK-I antibodies can react specifically with human IKK epsilon/IKK-I and show no cross reactivity with IKK α, IKK β and IKK γ.

Specificity

The antibody show no cross-reactivity to IKKα, IKKβ, or IKKγ.

Immunogen

Synthetic peptide corresponding to amino acids 701-716 of human IKK-epsilon/IKK-I conjugated to KLH.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Polyclonal anti-IKK epsilon/IKK-I c-terminal antibody can be used in immunoprecipitation and western blot.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Ping Guo et al.
The Journal of biological chemistry, 285(6), 3676-3684 (2009-11-27)
IKKepsilon has recently been identified as a breast cancer oncogene. Elevated levels of IKKepsilon are associated with cell survival and growth. Here, we show that IKKepsilon interacts with and phosphorylates estrogen receptor alpha (ERalpha) on serine 167 in vitro and
Suhu Liu et al.
Blood advances, 2(23), 3428-3442 (2018-12-07)
To identify novel therapeutic targets in acute myeloid leukemia (AML), we examined kinase expression patterns in primary AML samples. We found that the serine/threonine kinase IKBKE, a noncanonical IkB kinase, is expressed at higher levels in myeloid leukemia cells compared
Israel Cañadas et al.
Nature medicine, 24(8), 1143-1150 (2018-07-25)
Mesenchymal tumor subpopulations secrete pro-tumorigenic cytokines and promote treatment resistance1-4. This phenomenon has been implicated in chemorefractory small cell lung cancer and resistance to targeted therapies5-8, but remains incompletely defined. Here, we identify a subclass of endogenous retroviruses (ERVs) that
T Shimada et al.
International immunology, 11(8), 1357-1362 (1999-07-28)
Using the suppression subtractive hybridization technique, we isolated a novel kinase, IKK-i, whose message is drastically induced by lipopolysaccharide (LPS) in the mouse macrophage cell line RAW264. 7. The predicted protein contains the kinase domain in its N-terminus, which shares
Serkan Ismail Göktuna
EXCLI journal, 21, 436-453 (2022-04-09)
IKBKE have been associated with numerous cancers. As a result, IKBKE have emerged as potential target for cancer therapy. Accumulating evidence support that IKBKE orchestrate tumor cell survival in cancers. Here we evaluated the possible link between IKBKE and ERK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service