Skip to Content
Merck
All Photos(1)

Key Documents

I4638

Sigma-Aldrich

Interleukin-1β Fragment 163-171 human

≥97% (HPLC), synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H64N12O19
CAS Number:
Molecular Weight:
1004.99
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

biological source

synthetic

Assay

≥97% (HPLC)

form

powder

mol wt

Mw 1004.99

packaging

pkg of 1 mg

color

white

solubility

water: soluble 1 mg/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C39H64N12O19/c1-17(2)31(43)38(68)47-18(6-9-25(41)53)32(62)44-15-27(55)45-19(7-10-28(56)57)33(63)46-20(8-11-29(58)59)34(64)51-24(16-52)37(67)49-22(13-26(42)54)35(65)50-23(14-30(60)61)36(66)48-21(39(69)70)5-3-4-12-40/h17-24,31,52H,3-16,40,43H2,1-2H3,(H2,41,53)(H2,42,54)(H,44,62)(H,45,55)(H,46,63)(H,47,68)(H,48,66)(H,49,67)(H,50,65)(H,51,64)(H,56,57)(H,58,59)(H,60,61)(H,69,70)/t18-,19-,20-,21-,22-,23-,24-,31-/m0/s1

InChI key

NIXBZDOQHRZFDR-OEWCNXHOSA-N

Gene Information

human ... IL1B(3553)

Amino Acid Sequence

Val-Gln-Gly-Glu-Glu-Ser-Asn-Asp-Lys

Biochem/physiol Actions

IL1B (interleukin 1 β) is an immune-activating cytokine. In presence of infection or cell stress, it gets converted into active mature form.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gulden Avci et al.
The Journal of surgical research, 172(1), e39-e46 (2011-11-15)
Curcumin has been shown to decrease ischemia-reperfusion (I/R) injury in kidney or brain tissues. In this study, the effects of curcumin were evaluated in skeletal muscle during I/R injury. Hind limb ischemia was induced by clamping the common femoral artery
G Antoni et al.
Journal of immunology (Baltimore, Md. : 1950), 137(10), 3201-3204 (1986-11-15)
Short peptide fragments of human and murine interleukin 1 (IL 1) were synthesized on the basis of their predicted exposure on the surface of the molecule in an attempt to identify the minimal structure responsible for the immunostimulatory activity of
Anna K Blakney et al.
Journal of biomedical materials research. Part A, 100(6), 1375-1386 (2012-03-13)
Poly(ethylene glycol) (PEG) hydrogels, modified with RGD, are promising platforms for cell encapsulation and tissue engineering. While these hydrogels offer tunable mechanical properties, the extent of the host response may limit their in vivo applicability. The overall objective was to
L Nencioni et al.
Journal of immunology (Baltimore, Md. : 1950), 139(3), 800-804 (1987-08-01)
The stimulating effect of a synthetic nonapeptide (fragment 163-171) of human interleukin 1 beta (IL-1 beta) on antibody responses to both T helper-dependent and T helper-independent antigens was investigated. It was shown that the nonapeptide enhanced the antibody response, as
Xianwei Wang et al.
Apoptosis : an international journal on programmed cell death, 17(8), 908-915 (2012-04-25)
Angiotensin II (Ang II) exerts its effects by activating its receptors, primarily type 1 (AT1R) and type 2 (AT2R). While the role of AT1R activation in cardiomyocyte physiology is well known, the role of AT2R in cardiomyocyte apoptosis remains controversial.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service