Skip to Content
Merck
All Photos(1)

Key Documents

I4259

Sigma-Aldrich

Iodoethane

Synonym(s):

Ethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5I
CAS Number:
Molecular Weight:
155.97
Beilstein:
505934
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

5.38 (vs air)

vapor pressure

100 mmHg ( 18 °C)

refractive index

n20/D 1.513 (lit.)

bp

69-73 °C (lit.)

mp

−108 °C (lit.)

density

1.940 g/mL at 25 °C (lit.)

SMILES string

CCI

InChI

1S/C2H5I/c1-2-3/h2H2,1H3

InChI key

HVTICUPFWKNHNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup - (External MSDS)

Flash Point(C)

53 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service