Skip to Content
Merck

Skip To

I3036

Polyinosinic–polycytidylic acid potassium salt

γ-irradiated, 10% Poly (I:C) with buffer salts, TLR ligand tested

Synonym(s):

Poly (I:C), Poly(I) • Poly(C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 mg
Check Cart for Availability
€ 138,00
20 mg
Check Cart for Availability
€ 714,00

About This Item

CAS Number:
UNSPSC Code:
41106305
NACRES:
NA.51
MDL number:
Assay:
(Free nucleotides NMT 1% (TLC))

€ 138,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

(Free nucleotides NMT 1% (TLC))

composition

, 8-12% UV

storage temp.

−20°C

Biochem/physiol Actions

Transfection of Poly (I:C) into NIT-1 cells has been used as a model of intracellular dsRNA-induced β cell apoptosis. Eighteen hours post transfection, 45% of the cells were apoptotic with an increase in NF-kB, p50/p65 nuclear translocation, and cleavage of caspases 3 and 8, as well as transcriptional induction of caspase 12, Fas, IL-15, and the TNF receptor-associated ligand (TRAIL). It has been suggested that Poly(I:C) is one of the most appropriate generators of stable mature dendritic cells (DC). These mature DC might generate in vivo effective immune responses after injection due to their ability to secrete bioactive IL-12 after CD40 ligation. Poly (I:C) was used as a potent adjuvant to enhance the specific anti-tumor immune responses against a peptide-based vaccine.

Packaging

Package size based on polynucleotide content

Physical form

10% Poly (I:C) with sodium chloride and sodium phosphate buffer salts

Other Notes

Double-stranded homopolymer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9582P4154P1530
assay

(Free nucleotides NMT 1% (TLC))

assay

(Free nucleotides NMT 1% (TLC))

assay

≥98% (TLC)

assay

(Free nucleotides NMT 1% (TLC)), ≥99% (less than 1% free nucleotides, TLC)

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

300

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

composition

, 8-12% UV

composition

Poly(I) • Poly(C), 10% (balance buffer salts as sodium chloride and sodium phosphate)

composition

-

composition

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters