Skip to Content
Merck
All Photos(1)

Key Documents

I2785

Sigma-Aldrich

IRAK4, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

IPD1, NY-REN-64, REN64

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€401.00

€401.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€401.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

€401.00


Estimated to ship on16 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

378-512 nmol/min·mg

mol wt

~81 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IRAK4(51135)

Biochem/physiol Actions

IRAK4 (interleukin-1 receptor-associated kinase 4) protein contains a death domain and a kinase-like domain. IRAKs are mainly involved in toll-like receptors (TLRs) and interleukin-1 receptors (IL-1Rs) signaling. IRAK4 can activate NF-κB (nuclear factor κ B) and MAPK (mitogen activated protein kinase) pathways. It plays a crucial role in adaptive immunity and inflammatory responses.[1][2][3] IRAK4 mRNA levels are higher in humans suffering from Vogt-Koyanagi-Harada (VKH) disease.[1] Mutations in IRAK4 are associated with recurrent pyogenic bacterial infections in children.[4]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan Ferrao et al.
Molecular cell, 55(6), 891-903 (2014-09-10)
Trans-autophosphorylation is among the most prevalent means of protein kinase activation, yet its molecular basis is poorly defined. In Toll-like receptor and interleukin-1 receptor signaling pathways, the kinase IRAK4 is recruited to the membrane-proximal adaptor MyD88 through death domain (DD)
Min Sun et al.
PloS one, 9(4), e93214-e93214 (2014-04-03)
To explore whether IRAK1 and IRAK4 are involved in the pathogenesis of Vogt-Koyanagi-Harada (VKH) disease. Thirty-nine VKH patients and thirty-two healthy controls were included in this study. The mRNA levels of IRAK1 and IRAK4 from active VKH patients, inactive VKH
Shyun Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(8), 5567-5572 (2002-04-18)
Toll/IL-1 receptor family members are central components of host defense mechanisms in a variety of species. One well conserved element in their signal transduction is Ser/Thr kinases, which couple early signaling events in a receptor complex at the plasma membrane
Nobutaka Suzuki et al.
Trends in immunology, 23(10), 503-506 (2002-09-26)
Toll-like receptors (TLRs), which recognize pathogen-associated molecular patterns and members of the proinflammatory interleukin-1 receptor (IL-1R) family, share homologies in their cytoplasmic domains. Engagement of members of both of these families initiates a common intracellular signaling cascade, in which MyD88
Leah Cushing et al.
The Journal of biological chemistry, 289(15), 10865-10875 (2014-02-26)
IRAK4 is a central kinase in innate immunity, but the role of its kinase activity is controversial. The mechanism of activation for IRAK4 is currently unknown, and little is known about the role of IRAK4 kinase in cytokine production, particularly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service