Skip to Content
Merck

Skip To

I2276

Interleukin-12 human

≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

IL-12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 μg
Check Cart for Availability
€ 953,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:

€ 953,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

assay

≥97% (SDS-PAGE and N-terminal analysis)

form

lyophilized powder

potency

0.01-0.05 ng/mL ED50

mol wt

glycoprotein 75 kDa

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Interleukin-12 (IL-12) was identified as a factor secreted by human Epstein-Barr (EBV)-transformed B cell lines. IL-12, or Natural Killer Cell Stimulatory Factor (NKSF), is a 75 kDa disulfide-linked heterodimer of a 35 kDa subunit and 40 kDa subunit. IL-12 is produced predominantly by monocytes and NK cells and induces T cells and NK cells to produce IFN-γ. Human IL-12 is not active on mouse cells, but murine IL-12 is active on both murine and human lymphocytes. The biological activity of recombinant human IL-12 was measured by its ability to stimulate the proliferation of PHA-activated human T lymphoblasts. IL-12 has anti-angiogenic properties.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H7666I1771I9276
biological source

human

biological source

human

biological source

human

biological source

human

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in E. coli

recombinant

expressed in baculovirus infected Sf21 cells

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

assay

≥97% (SDS-PAGE and N-terminal analysis)

assay

≥95% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized

form

lyophilized powder

form

lyophilized powder


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters