Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
2 mL | Check Cart for Availability | € 216,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
indirect ELISA: 1:80,000, quantitative precipitin assay: 3.0-4.0 mg/mL
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 200 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source goat |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form IgG fraction of antiserum | antibody form IgG fraction of antiserum | antibody form whole antiserum | antibody form IgG fraction of antiserum |
| form buffered aqueous solution | form buffered aqueous solution | form - | form buffered aqueous solution |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Still not finding the right product?
Explore all of our products under Anti-Human IgG (whole molecule) antibody produced in rabbit
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



