Skip to Content
Merck
All Photos(1)

Key Documents

I122

Sigma-Aldrich

ICI 204,448 hydrochloride

solid

Synonym(s):

(±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride, R,S-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl] phenoxy]-acetic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26Cl2N2O4 · HCl
CAS Number:
Molecular Weight:
501.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 12 mg/mL
dilute sodium carbonate: 4 mg/mL
H2O: 4.5 mg/mL

SMILES string

Cl[H].CN(C(CN1CCCC1)c2cccc(OCC(O)=O)c2)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C23H26Cl2N2O4.ClH/c1-26(22(28)12-16-7-8-19(24)20(25)11-16)21(14-27-9-2-3-10-27)17-5-4-6-18(13-17)31-15-23(29)30;/h4-8,11,13,21H,2-3,9-10,12,14-15H2,1H3,(H,29,30);1H

InChI key

FNDGLVOYAQNQPE-UHFFFAOYSA-N

Gene Information

human ... OPRK1(4986)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

κ opioid receptor agonist that does not cross the blood-brain barrier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Keïta et al.
European journal of pharmacology, 277(2-3), 275-280 (1995-04-24)
The antinociceptive effect of intraplantar (i.pl.) ICI 204448 ((R,S)-N-[2-(N-methyl-3,4-dichloro-phenylacetamido)-2-(3-carbox yph enyl)- ethyl]pyrrolidine hydrochloride)) (20, 30, 40 and 50 micrograms), a kappa-opioid receptor agonist which has limited access to the central nervous system, was studied in a well-established rat model of
R A Hirst et al.
Neuroscience letters, 232(2), 119-122 (1997-08-29)
This study describes the coupling of the recombinant rat kappa-opioid receptor expressed in Chinese hamster ovary (CHO) cells to adenylyl cyclase and the effects of receptor density. The binding of [3H]diprenorphine ([3H]DPN) was dose dependent and saturable in membranes prepared
Scott M Rawls et al.
Pharmacology, 74(2), 79-83 (2005-02-03)
ICI 204448, a selective kappa-opioid agonist with limited CNS access, can be used to discriminate central and peripheral opioid actions on physiological systems such as pain and thermoregulation. Therefore, we investigated the effect of ICI 204448 (2.5, 5, and 10
E R Butelman et al.
Psychopharmacology, 147(1), 73-80 (1999-12-11)
The dynorphins are endogenous opioid peptides with relative binding selectivity for kappa-receptors. It is unclear whether the dynorphins share the pharmacological profile observed with synthetic kappa-agonists in primates. The main objective of this study was to compare the effects of
A Barber et al.
British journal of pharmacology, 113(4), 1317-1327 (1994-12-01)
1. The pharmacological properties of the novel diarylacetamide kappa-opioid receptor agonist, EMD 61753, have been compared with those of ICI 197067 (a centrally-acting kappa agonist) and ICI 204448 (a peripherally-selective kappa agonist). 2. EMD 61753 binds with high affinity (IC50

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service