Skip to Content
Merck
All Photos(1)

Key Documents

I0406

Sigma-Aldrich

IL-6 rat

>97% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture

Synonym(s):

Interleukin-6 from rat

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€310.00
50 μG
€1,090.00

€310.00


Check Cart for Availability


Select a Size

Change View
10 μG
€310.00
50 μG
€1,090.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€310.00


Check Cart for Availability

Product Name

Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

buffered aqueous solution

potency

0.01-0.06 ng/mL ED50

quality

endotoxin tested

mol wt

predicted mol wt ~22 kDa

packaging

pkg of 10 and 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.01EU/mg (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... Il6(24498)

Biochem/physiol Actions

Interleukin-6 (IL-6) is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 induces multiple effects, as indicated by its numerous synonyms: plasmacytoma growth factor (PCT-GF), interferon-β-2 (IFN-β2), monocyte derived human B cell growth factor, B cell stimulating factor (BSF-2), hepatocyte stimulating factor (HSF), Interleukin Hybridoma/Plasmacytoma-1 (IL-HP1). In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 also potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.

Physical form

Supplied as a 0.2 μm filtered solution in 50 mM sodium acetate, pH 4, with BSA as a carrier protein.

Preparation Note

Recombinant, rat Interleukin-6 is produced from a DNA sequence encoding the mature rat IL-6 protein. The methionyl form of mature rat IL-6, a 188 amino acid protein, has a predicted molecular mass of approx. 22 kDa. Rat and mouse IL-6 share 93% amino acid sequence identity.

Analysis Note

The biological activity is measured in a cell proliferation assay using IL-6 dependent mouse plasmacytoma cell line, T1165.85.2.1.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P W Bodell et al.
Journal of applied physiology (Bethesda, Md. : 1985), 106(2), 443-453 (2008-12-06)
Childhood diseases are often accompanied by chronic inflammation, which is thought to negatively impact growth. Interleukin-6 (IL-6) is typically cited as an indicator of inflammation and is linked to impaired growth. This study was designed to isolate and identify potential
Elin Banke et al.
Metabolism: clinical and experimental, 63(6), 831-840 (2014-04-02)
The bacteria Staphylococcus aureus is part of the normal bacterial flora and produces a repertoire of enterotoxins which can cause food poisoning and toxic shock and might contribute to the pathogenesis of inflammatory diseases. These enterotoxins directly cross-link the T
Lorena M Amaral et al.
American journal of obstetrics and gynecology, 211(2), 158-158 (2014-02-20)
Preeclampsia is a multisystem disorder recognized as hypertension with proteinuria developing >20 weeks' gestation. Preeclampsia is associated with chronic immune activation characterized by increased T and B lymphocytes, cytokines, and antibodies activating the angiotensin II type I receptor (AT1-AA). Hypertension
Mingming Zhang et al.
PloS one, 9(10), e109146-e109146 (2014-10-03)
It has been shown that Faecalibacterium prausnitzii (F. prausnitzii), one of the dominant intestinal bacterial flora, may protect colonic mucosa against the development of inflammation and subsequent inflammatory bowel disease (IBD), with the underlying mechanisms being unclear. The impacts of
A O'Toole et al.
British journal of cancer, 111(5), 927-932 (2014-07-25)
Tumour microenvironment (TME) of advanced colorectal cancer (CRC) suppresses dendritic cell (DC) maturation. Here, our aim was to determine how the microenvironment of early-stage tumours influences DCs. Tumour-conditioned media (TCM) was generated by culturing explant tumour tissue in vitro (n=50).

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service