I0375
4-Imidazoleacetic acid hydrochloride
Synonym(s):
(4-Imidazolyl)acetic acid hydrochloride, I4AA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
mp
218-222 °C (lit.)
storage temp.
2-8°C
SMILES string
Cl.OC(=O)Cc1c[nH]cn1
InChI
1S/C5H6N2O2.ClH/c8-5(9)1-4-2-6-3-7-4;/h2-3H,1H2,(H,6,7)(H,8,9);1H
InChI key
MWHLCFYPFGFBQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Competitive antagonist at GABAC receptors.
replaced by
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
188.6 °F - Solvent
Flash Point(C)
87 °C - Solvent
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service