Skip to Content
Merck

Skip To

HT40232

Papanicolaou Stain, Modified EA 65

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 L
Check Cart for Availability
€ 89,80

About This Item

UNSPSC Code:
41116121
NACRES:
NA.47
MDL number:

€ 89,80


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HT40432HT40316HT40332
form

solution

form

solution

form

solution

form

solution

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

General description

For routine diagnostic cytology to aid in the identification and classification of exfoliative cells.

Application

Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.

Other Notes

Eosin Y (certified), 0.25% w/v,fast green FCF (certified), 0.004% w/v, and phosphotungstic acid, 0.4% w/v, in denatured alcohol with stabilizer

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Kurzinformation zum Gebiet der Zytologie, ihrer Rolle bei der Zytodiagnostik und zum Einsatz von Standard- und Spezialfärbungen für die mikroskopische Analyse zum Nachweis von Zellanomalien.

Global Trade Item Number

SKUGTIN
HT40232-1L04061838207661

Questions

  1. I want to perform conjunctival impression cytology, is Papanicolaou Stain Modified EA 65 appropriate if I want to fix and stain specimens with a combination of periodic acid-Schiff (PAS) and Gill’s modified Papanicolaou stains?

    1 answer
    1. This product has not been tested for its suitability in dual staining. When using the PAP Modified EA65 stain, the slides are also routinely stained with Gill's hematoxylin and OG-6. Using just the Papanicolaou Stain Modified EA 65 with a PAS stain would be considered "off-label" use by the FDA, as the PAP stain reagents are approved by the Food and Drug Administration for use in performing a standard PAP stain procedure.
      This application will need to be validated and properly documented for staining clinical specimens by the end-user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service