Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
2.5 L | Check Cart for Availability | € 232,00 |
About This Item
form
solution
Quality Segment
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
concentration
0.5 % (w/v) in water
application(s)
hematology
histology
storage temp.
room temp
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| concentration 0.5 % (w/v) in water | concentration 0.5 % (w/v) in water | concentration 0.5 % (w/v) in water | concentration 0.5 % (w/v) in water |
| form solution | form solution | form solution | form solution |
| application(s) hematology | application(s) hematology | application(s) hematology | application(s) hematology |
| IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. |
Still not finding the right product?
Explore all of our products under Eosin Y Solution, Aqueous
Storage Class
10 - Combustible liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
