Skip to Content
Merck
All Photos(2)

Documents

HT101A

Sigma-Aldrich

Silver Stain Kit, modified Steiner-Steiner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label

application(s)

hematology
histology

storage temp.

2-8°C

Application

Test procedure includes both standard and microwave applications.

Diagnostic Implications

Useful in identifying spirochetes and nonfilamentous bacteria.

Kit Components Only

Product No.
Description

  • Gum Mastic Solution (kit only) 500 mL

  • Hydroquinone Tablets (HT1014-10TAB) 10 tablets

  • Silver Nitrate Solution (kit only) 500 mL

  • Uranyl Nitrate Solution (kit only) 500 mL

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

66.0 °F

Flash Point(C)

18.88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda G Vincent et al.
Diseases of aquatic organisms, 67(1-2), 163-169 (2006-01-03)
Necrotizing hepatopancreatitis (NHP), a severe bacterial disease affecting penaeid shrimp aquaculture, is caused by a gram-negative, pleomorphic, intracellular alpha-proteobacterium referred to as the NHP-bacterium (NHPB). The time course of NHP was investigated in experimentally infected juveniles of Kona stock Litopenaeus

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service