Merck
Skip To
HS23401
1-Place Compartment Dispensing Bin
clear (bin)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 81,00
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
€ 81,00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
acrylic , clear (bin)
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23401
L × W × H
184 mm × 194 mm × 232 mm
General description
The Workstation Dispensing Bin′s crystal-clear bins let you see your small items inventory.
Features and Benefits
  • One Compartment
  • See inventory levels of pipette tips, tubes, fittings, finger cots, and other bulk small items at a glance
  • Flip-top covers on generous openings make filling and dispensing easy and keep products dust-free
  • Includes non-skid rubber feet
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
HS23402HS23403HS23404
description
clear (bin)
description
clear (bin)
description
clear (bin)
description
clear (bin)
manufacturer/tradename
Heathrow Scientific HS23401
manufacturer/tradename
Heathrow Scientific HS23402
manufacturer/tradename
Heathrow Scientific HS23403
manufacturer/tradename
Heathrow Scientific HS23404
L × W × H
184 mm × 194 mm × 232 mm
L × W × H
150 mm × 177 mm × 368 mm
L × W × H
217 mm × 170 mm × 217 mm
L × W × H
287 mm × 170 mm × 217 mm
material
acrylic , clear (bin)
material
acrylic , clear (bin)
material
acrylic , clear (bin)
material
acrylic , clear (bin)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
Choose from one of the most recent versions:
Certificates of Analysis (COA)