Key Documents
HS120554
Pipette Starter Kit
single-channel
Select a Size
1 EACH
€429.00
€429.00
Check Cart for Availability
Select a Size
Change View
1 EACH
€429.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€429.00
Check Cart for Availability
feature
adjustable
single-channel
manufacturer/tradename
Heathrow Scientific 120554
volume × volume × volume
2-20 μL × 20-200 μL × 100-1000 μL
Related Categories
General description
The Pipette Starter Kit is convenient for new labs or for customers who want a new set of pipettors covering the most commonly used volumes. The kit includes 3 single channel Pearl Pipettes, a Universal Carousel Pipette Stand and five Reagent Reservoirs–all at a value price!
Features and Benefits
Universal Carousel Pipette Stand (120499):
  • Holds up to eight single-channel, four multi-channel, or a combo of both pipettes
  • Weighted base allows for easy removal or replacement of pipettes, with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • Assorted color options allow for color coding pipette location
  • The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
  • U.S. Patent Pending
Pearl Pipettes (HS120104 100-1000 μL, HS120102 20-200μL, & HS120099 2-20μL)
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digital volume display
  • Labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified
  • Autoclavable
Reagent Reservoirs: (120644)
  • Tapered reservoir bottoms form into a narrow trough to aid full sample recovery
  • Disposable
  • Sterile
Legal Information
Pearl is a trademark of Specac, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)