HS120316
TrueNorth® Microscope Slide Box
25 Place, Fitted Lid, blue polycarbonate
All Photos(1)
About This Item
Recommended Products
material
blue polycarbonate
feature
Holds 25 x slides 76x26 mm
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120316
L × W × H
141 × 88 × 35 mm
Features and Benefits
- protect your slides in ultra low temperature ranges down to -80C
- durable polycarbonate and stainless steel clasp/friction fit lid
- safe storage and transport of 25 x 75 mm slides (3 x 1 in.)
- uniform grooved slide slots separate slides preventing slide contact
- temperature-resistant numbered foam lining and identification sheet
- stackable
- autoclavable
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC