Skip to Content
Merck
All Photos(7)

Key Documents

HPA047192

Sigma-Aldrich

Anti-CLCA2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CLCRG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EELTLSWTAPGEDFDQGQATSYEIRMSKSLQNIQDDFNNAILVNTSKRNPQQAGIREIFTFSPQISTNGPEHQPNGETHESHRIYVAIRAMDRNSLQSAVSNIAQAPLFIPPNSDPVPARDYLILKGVLTAMGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLCA2(9635)

General description

Calcium activated chloride channel 2(CLCA2) is an integral transmembrane protein. CLCA2 is induced during human mammary epithelial cell differentiation. CLCA2 is located on human chromosome 1p22.3.

Immunogen

chloride channel accessory 2

Application

Anti-CLCA2 antibody has been used to detect CLCA2 in nasopharyngeal carcinoma(NPC) cell lines. It also has been used to detect CLCA2 in SiHa cells and tumor tissues using western blotting.

Biochem/physiol Actions

CLCA2 regulates cell migration and invasion and is also associated with p53 tumour suppressor genes. CCLA2 helps to maintain corneal transparency due to the high and tissue specific expression of CCLA2 gene in human corneal epithelium. CLCA2 plays an important role in suppressing breast cancer. CLCA2 is used as an effective immunohistochemical marker for distinguishing primary SCC and primary ADC of lungs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71988

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CLCA2, a novel RUNX1 partner gene in a therapy-related leukemia with t (1; 21)(p22; q22)
Giguere A, et al.
Cancer Genetics and Cytogenetics, 202(2), 94-100 (2010)
Peijin Zhang et al.
Experimental and therapeutic medicine, 22(1), 776-776 (2021-06-01)
Ca2+-activated Cl- channel A2 (CLCA2), a tumor suppressor, is associated with the development of several cancers. However, little is known about CLCA2 in human cervical cancer. Therefore, the aim of the present study was to investigate the effects of CLCA2
CLCA2 interactor EVA1 is required for mammary epithelial cell differentiation
Ramena, G, et al.
PLoS ONE, 11(3), e0147489-e0147489 (2016)
CLCA2 tumour suppressor gene in 1p31 is epigenetically regulated in breast cancer
Li, Xiuro, et al.
Oncogene, 23(7), 1474-1474 (2004)
CLCA2 as a novel immunohistochemical marker for differential diagnosis of squamous cell carcinoma from adenocarcinoma of the lung
Shinmura,, et al.
Disease Markers, 2014 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service